Learn More
Medchemexpress LLC Ribostamycin | 25546-65-0 | 99.89% | C17H34N4O10 | 25 MG
SDP
Catalog No. 5000356085
Encompass_Preferred
Ribostamycin (Vistamycin) is a broad-spectrum aminoglycoside antibiotic. It is effective against Gram-Negative and Gram-Positive bacterial infections, and it also inhibits the chaperone activity of PDI.
  • Broad-spectrum aminoglycoside antibiotic
  • Effective against Gram-Negative and Gram-Positive bacterial infections
  • Inhibits the chaperone activity of PDI
  • Inhibits *Borrelia burgdorferi*
  • Inhibits *Escherichia coli* strains
Catalog No. 50-003-56085 Supplier Medchemexpress LLC Supplier No. 25MGHY142127
Ribostamycin (Vistamycin) is a broad-spectrum aminoglycoside antibiotic. It is effective against Gram-Negative and Gram-Positive bacterial infections, and it also inhibits the chaperone activity of PDI.
  • Broad-spectrum aminoglycoside antibiotic
  • Effective against Gram-Negative and Gram-Positive bacterial infections
  • Inhibits the chaperone activity of PDI
  • Inhibits *Borrelia burgdorferi*
  • Inhibits *Escherichia coli* strains