Learn More
Medchemexpress LLC Ribostamycin | 25546-65-0 | 99.9% | C17H34N4O10 | 50 MG
SDP
Catalog No. 5000353691
Encompass_Preferred
Ribostamycin is a broad-spectrum aminoglycoside antibiotic effective against Gram-Negative and Gram-Positive bacterial infections. It also acts as an inhibitor of PDI chaperone activity.
  • Broad-spectrum aminoglycoside antibiotic
  • Effective against Gram-Negative and Gram-Positive bacterial infections
  • Inhibits chaperone activity of PDI
  • Targets bacterial processes and acts as an antibiotic
  • Involved in anti-infection pathways
  • Demonstrates in vitro inhibition of Borrelia burgdorferi and Escherichia coli
  • Shows minimal nephrotoxicity in in vivo studies
Catalog No. 50-003-53691 Supplier Medchemexpress LLC Supplier No. HY14212750MG
Ribostamycin is a broad-spectrum aminoglycoside antibiotic effective against Gram-Negative and Gram-Positive bacterial infections. It also acts as an inhibitor of PDI chaperone activity.
  • Broad-spectrum aminoglycoside antibiotic
  • Effective against Gram-Negative and Gram-Positive bacterial infections
  • Inhibits chaperone activity of PDI
  • Targets bacterial processes and acts as an antibiotic
  • Involved in anti-infection pathways
  • Demonstrates in vitro inhibition of Borrelia burgdorferi and Escherichia coli
  • Shows minimal nephrotoxicity in in vivo studies