Learn More
TARGETMOL CHEMICALS INC Ribostamycin sulfate 200MG
SDP
Catalog No. 502878349
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ribostamycin sulfate (Vistamycin sulfate) , an aminoglycoside antibiotic, contains a neutral sugar moiety and is produced by Streptomyces ribosome. Purity 99.72%
Catalog No. 50-287-8349 Supplier TARGETMOL CHEMICALS INC Supplier No. T1214200MG
Also available in 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ribostamycin sulfate (Vistamycin sulfate) , an aminoglycoside antibiotic, contains a neutral sugar moiety and is produced by Streptomyces ribosome. Purity 99.72%