Learn More
Apexbio Technology LLC Ribostamycin Sulfate 53797-35-6 10mM (in 1mL H20)
SDP
Catalog No. 5000090206
Encompass_Preferred
Ribostamycin Sulfate (CAS 53797-35-6) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to disrupt bacterial protein synthesis thereby inhibiting bacterial growth Ribostamycin Sulfate exerts its biological activity primarily through binding to the 30S ribosomal subunit and impairing protein translation In in vitro assays Ribostamycin typically demonstrates inhibitory activity within the micromolar concentration range though specific IC50 values vary depending on bacterial species and experimental conditions Based on these pharmacological properties Ribostamycin Sulfate holds research potential in the study of bacterial protein translation mechanisms evaluation of antibiotic resistance phenotypes and antimicrobial susceptibility testing in microbial strains
Catalog No. 50-000-90206 Supplier Apexbio Technology LLC Supplier No. B18255.1
Ribostamycin Sulfate (CAS 53797-35-6) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to disrupt bacterial protein synthesis thereby inhibiting bacterial growth Ribostamycin Sulfate exerts its biological activity primarily through binding to the 30S ribosomal subunit and impairing protein translation In in vitro assays Ribostamycin typically demonstrates inhibitory activity within the micromolar concentration range though specific IC50 values vary depending on bacterial species and experimental conditions Based on these pharmacological properties Ribostamycin Sulfate holds research potential in the study of bacterial protein translation mechanisms evaluation of antibiotic resistance phenotypes and antimicrobial susceptibility testing in microbial strains