Learn More
Medchemexpress LLC Ribostamycin sulfate | 53797-35-6 | 99.5% | 25 MG
SDP
Catalog No. 5000414202
Encompass_Preferred
Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial that inhibits bacterial protein synthesis. It also inhibits the chaperone activity of protein disulfide isomerase (PDI) and is used in pharmacokinetic and nephrotoxicity studies.
  • Functions as a broad-spectrum antimicrobial.
  • Inhibits bacterial protein synthesis at the 30S and 50S ribosomal subunits.
  • Inhibits chaperone activity of protein disulfide isomerase (PDI).
  • Suitable for use in pharmacokinetic and nephrotoxicity studies.
Catalog No. 50-004-14202 Supplier Medchemexpress LLC Supplier No. 25MGHYB1228
Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial that inhibits bacterial protein synthesis. It also inhibits the chaperone activity of protein disulfide isomerase (PDI) and is used in pharmacokinetic and nephrotoxicity studies.
  • Functions as a broad-spectrum antimicrobial.
  • Inhibits bacterial protein synthesis at the 30S and 50S ribosomal subunits.
  • Inhibits chaperone activity of protein disulfide isomerase (PDI).
  • Suitable for use in pharmacokinetic and nephrotoxicity studies.