Learn More
Medchemexpress LLC Ribostamycin sulfate | 53797-35-6 | 99.55% | 100 MG
SDP
Catalog No. 5000412991
Encompass_Preferred
Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial that inhibits bacterial protein synthesis at the level of 30S and 50S ribosomal subunit binding. It also inhibits the chaperone activity of protein disulfide isomerase (PDI). It is used in pharmacokinetic and nephrotoxicity studies.
  • Broad-spectrum antimicrobial activity.
  • Inhibits bacterial protein synthesis at 30S and 50S ribosomal subunits.
  • Inhibits the chaperone activity of protein disulfide isomerase (PDI).
  • Effective against both gram-positive and gram-negative cocci and bacilli, including drug-resistant strains.
  • Especially effective against gentamicin-resistant *Klebsiella pneumoniae*.
  • Available for research use only.
Catalog No. 50-004-12991 Supplier Medchemexpress LLC Supplier No. HYB1228100MG
Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial that inhibits bacterial protein synthesis at the level of 30S and 50S ribosomal subunit binding. It also inhibits the chaperone activity of protein disulfide isomerase (PDI). It is used in pharmacokinetic and nephrotoxicity studies.
  • Broad-spectrum antimicrobial activity.
  • Inhibits bacterial protein synthesis at 30S and 50S ribosomal subunits.
  • Inhibits the chaperone activity of protein disulfide isomerase (PDI).
  • Effective against both gram-positive and gram-negative cocci and bacilli, including drug-resistant strains.
  • Especially effective against gentamicin-resistant *Klebsiella pneumoniae*.
  • Available for research use only.