Learn More
Medchemexpress LLC Ribostamycin sulfate | 53797-35-6 | MFCD23380549 | 10 MG
SDP
Catalog No. 5000298201
Encompass_Preferred
Ribostamycin sulfate is an aminoglycoside antibiotic (sulfate salt) used as a broad-spectrum antimicrobial that inhibits bacterial protein synthesis by binding to the 30S and 50S ribosomal subunits. It is used in antimicrobial research, pharmacokinetic studies, and nephrotoxicity studies.
  • Broad-spectrum antimicrobial activity against Gram-positive and Gram-negative bacteria.
  • Mechanism of action: inhibits bacterial protein synthesis at the ribosomal subunit level.
  • Sulfate salt form for enhanced handling and solubility in aqueous media.
  • Used in pharmacokinetic and nephrotoxicity research applications.
  • Well-characterized chemical identifiers (CAS and molecular weight available).
Catalog No. 50-002-98201 Supplier Medchemexpress LLC Supplier No. HYB122810MG
Ribostamycin sulfate is an aminoglycoside antibiotic (sulfate salt) used as a broad-spectrum antimicrobial that inhibits bacterial protein synthesis by binding to the 30S and 50S ribosomal subunits. It is used in antimicrobial research, pharmacokinetic studies, and nephrotoxicity studies.
  • Broad-spectrum antimicrobial activity against Gram-positive and Gram-negative bacteria.
  • Mechanism of action: inhibits bacterial protein synthesis at the ribosomal subunit level.
  • Sulfate salt form for enhanced handling and solubility in aqueous media.
  • Used in pharmacokinetic and nephrotoxicity research applications.
  • Well-characterized chemical identifiers (CAS and molecular weight available).