Learn More
Selleck Chemical LLC Ribostamycin Sulfate S4215-50mg
SDP
Catalog No. 503111217
Encompass_Preferred
Ribostamycin Sulfate(Vistamycin Sulfate Landamycine) is an aminoglycoside antibiotic containing a neutral sugar moiety and is produced by Streptomyces ribosidi cus
Catalog No. 50-311-1217 Supplier Selleck Chemical LLC Supplier No. S421550MG
Ribostamycin Sulfate(Vistamycin Sulfate Landamycine) is an aminoglycoside antibiotic containing a neutral sugar moiety and is produced by Streptomyces ribosidi cus