Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribostamycin Sulfate, G-Biosciences
SCI_ED

Catalog No. S139832
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139832 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139832 Supplier G-Biosciences Supplier No. RC1094
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribostamycin is an aminoglycoside antibiotic isolated from Streptomyces ribosifidicus. It is a broad-spectrum antibiotic targeting gram-positive and gram-negative bacteria.

Ribostamycin has been used to in pharmacokinetic studies and to study kidney toxicity. It is also an inhibitor of protein disulfide isomerase (PDI).

Chemical Identifiers

CAS 53797-35-6

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 250 mg
Percent Purity 95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.