Learn More
Aviva Systems Biology Corp RIC8A Antibody - N-terminal region (ARP57567_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57567_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIC8A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RIC8A. Alternate names: MGC104517, MGC131931, MGC148073, MGC148074, RIC8, synembrynApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-334