Learn More
Aviva Systems Biology Corp RIC8B Antibody - middle region (ARP57131_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIC8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RIC8B. Alternate names: FLJ10620, MGC39476, RIC8, hSynApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62088