Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gram's Iodine, Dilute Lugol's Iodine, Ricca Chemical
SDP

Catalog No. 345032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Volume (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
3450-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3450-32 Supplier Ricca Chemical Company Supplier No. 345032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gram's Iodine, Dilute Lugol's Iodine for gram staining.

  • For Gram Staining

Compliance

EPA (SW-846) listed as reagent for Total Coliform test with Multiple Tube Fermentation Technique. Meets ASTM standards as a reagent for Iron Bacteria test and Clostridium perfringens test.

Specifications

Quantity 1 L
Certifications/Compliance ASTM
For Use With (Application) Gram staining
Color Dark brown
Composition Iodine, Potassium iodide
Storage Requirements 15° to 30°C
Stain Type Iodine
Volume (Metric) 1 L
Sterility Non-sterile
Product Type Gram's Iodine
Packaging Amber Glass
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.