Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Davidson's Fixative, Modified, Ricca Chemical
SDP

Catalog No. R254490020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R254490020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R254490020F Supplier Ricca Chemical Company Supplier No. R254490020F
Add to Cart
Add to Cart

Specifications

Physical Form Solution
Chemical Name or Material Davidson's Fixative
CAS 7732-18-5, 64-17-5, 64-19-7, 50-00-0, 67-56-1, 67-63-0
Odor Slightly pungent
Packaging Cubitainer
Color Colorless
pH 2.5
Quantity 20 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.