Learn More
Epredia™ Parapel™ Paraffin Repellent
Catalog No. 22010277 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 oz. pump bottle; 6/CS
Catalog No. Quantity
22-010-277 4 oz. pump bottle; 6/CS
1 options
This item is not returnable. View return policy
Catalog No. 22-010-277 Supplier Epredia™ Supplier No. 2300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protect against excess buildup of wax on countertops and instrumentation with Epredia™ Parapel™ Paraffin Repellent.
  • Low odor
  • Reduces daily cleaning times
  • Will not leave behind oily residue
  • Will not remove finish from surfaces
Specifications
Quantity 4 oz. pump bottle; 6/CS