Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Richard-Allan Scientific™ Alcian Blue pH 2.5 stain solution

Catalog No. 21124395 Shop All Epredia Products
Change view
Click to view available options
ProductType: Alcian Blue pH 2.5 stain
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-124-395 ProductType: Alcian Blue pH 2.5 stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-395 Supplier Epredia™ Supplier No. 88043
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections.

  • Epredia histology and cytology stains produce consistent, clear results for hospitals, cancer centers and private labs
  • High-Performance
  • Achieve optimum staining performance
  • Nuclear clarity and superb definition
  • Wide variety of easy-to-use special stains

Specifications

Color Blue
pH 2.5
Product Type Alcian Blue pH 2.5 Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.