Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Catalog No. 41754 Shop All Epredia Products
Change view
Click to view available options
Capacity (Metric):
5.09 L
Catalog No. Capacity (Metric)
41-754 5.09 L
1 options
This item is not returnable. View return policy
Catalog No. 41-754 Supplier Epredia™ Supplier No. 41754
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Product Type Dry Specimen Container
Closure Included Yes
Material Polyethylene
Sterility Non-sterile
Capacity (Metric) 5.09 L
Closure Type Snap Cap
Color Opaque
No. per Case 10/Cs.