Learn More
Epredia™ Dry Specimen Containers - Bio-Tite™
Catalog No. 22110636 Shop All Epredia Products
Change view
Click to view available options
Capacity (Metric):
20 mL
Catalog No. Capacity (Metric)
22-110-636 20 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-110-636 Supplier Epredia™ Supplier No. 59202
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Specifications
Product Type Dry Specimen Container
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 20 mL
Closure Type Bio-Tite Lid
Color Opaque
No. per Case 384/Cs.