Learn More
Epredia™ Bouin's Fluid
GSA_VA
Catalog No. 22110621 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-110-621 1 gal.
1 options
Catalog No. 22-110-621 Supplier Epredia™ Supplier No. 57211
Add to Cart
Edge
Add to Cart
Preserve gastrointestinal tract specimens and structures with soft and delicate textures with Epredia™ Bouin's Fluid.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Capacity 3.79L
Product Type Fixative
Quantity 1 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov