Learn More
Epredia™ Richard-Allan Scientific™ Buffered Wright-Giemsa Stain
Catalog No. 22050271 Shop All Epredia Products
Change view
Click to view available options
Quantity:
32 oz.
Catalog No. Quantity
22-050-271 32 oz.
1 options
This item is not returnable. View return policy
Catalog No. 22-050-271 Supplier Epredia™ Supplier No. 3121
Add to Cart
Add to Cart
This item is not returnable. View return policy
A single solution stain that contains the fixative, buffer, and stain for rapid and convenient staining of blood smears.
  • Suitable for use on automated systems.
Specifications
Quantity 32 oz.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov