Learn More
Epredia™ Buffered Wright Stain
Catalog No. 3111 Shop All Epredia Products
Change view
Click to view available options
Quantity:
32 oz.
Catalog No. Quantity
3111 32 oz.
1 options
This item is not returnable. View return policy
Catalog No. 3111 Supplier Epredia™ Supplier No. 3111
This item is not returnable. View return policy
A single solution stain that contains fixative, buffer, and stain for rapid and convenient staining of blood smears.
Packaged in a 32 oz. bottle for ease of use; suitable for use on automated systems.
Specifications
Quantity 32 oz.
Description Buffered Wright Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov