Learn More
Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution
Shop All Thermo Scientific ProductsDescription
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, a mixture of formic acid and formaldehyde which allows simultaneous decalcification and fixation.
- Beneficial for conventional iron staining
- Preserves nuclear chromatin staining, even after prolonged exposure
- Eliminates poor hematoxylin and eosin staining caused by over-decalcification and/or exposure to harsh mineral acids.
Specifications
Specifications
| Product Type | Acetone |
| Quantity | 55 gal. |