Learn More
Epredia™ Cytocool™ II, Aerosol Freezing Spray
GSA_VA
Catalog No. 22050123 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cytocool II Aerosol Freezing Spray
Catalog No. Product Type
22-050-123 Cytocool II Aerosol Freezing Spray
1 options
This item is not returnable. View return policy
Catalog No. 22-050-123 Supplier Epredia™ Supplier No. 8323
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Flash freeze tissue specimens in the cryostat with Cytocool™ II.
  • Difficult-to-cut tissue can be hardened on microtome during sectioning
  • No ozone-depleting chlorofluorocarbons (CFCs)
  • Simple variation of finger pressure on the trigger nozzle controls tissue freezing; this feature helps to prevent over-cooling of specimen
  • Integral trigger nozzle eliminates need to attach separate handle to each can, making Cytocool II can less bulky and more comfortable to hold during use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Description 11 oz. can; 12/Cs
Product Type Cytocool II Aerosol Freezing Spray
WARNING: Reproductive Harm - www.P65Warnings.ca.gov