Learn More
Epredia™ Decalcifying Solution
Catalog No. 22050263 Shop All Epredia Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-263 1 gal.
22-050-130 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-050-263 Supplier Epredia™ Supplier No. 83401
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
  • Offers rapid decalcification without destroying nuclear properties of the tissue
  • The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
Specifications
Product Type Decalcifying Solution
Quantity 1 gal.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov