Learn More
Epredia™ Richard-Allan Scientific™ Differentiating Solution
Catalog No. 22110640 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-110-640 1 gal.
22-110-688 4 x 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-640 Supplier Epredia™ Supplier No. 74211
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Obtain clear differentiation and contrast between the hematoxylin and eosin with Epredia™ Richard-Allan Scientific™ Differentiating Solution.
  • Reproducible batch results allow for consistent staining in regressive procedures
Specifications
Product Type Hydrochloric Acid Solution
Quantity 1 gal.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov