Learn More
Epredia™ Richard Allan Scientific Dry Specimen Containers
Catalog No. 22046317 Shop All Epredia Products
Change view
Click to view available options
Capacity (Metric):
480 mL
Catalog No. Capacity (Metric)
22-046317 480 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-046317 Supplier Epredia™ Supplier No. 5316
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type Dry Specimen Container
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 480 mL
Closure Type Snap Cap
Color Opaque
No. per Case 350
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov