Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
Catalog No. 22110687 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
4 x 1 pt.
Product Type:
Alcoholic Eosin Y Stain
Eosin Phloxine Stain
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-110-687 4 x 1 pt. Alcoholic Eosin Y Stain
22-110-637 1 pt. Alcoholic Eosin Y Stain
22-110-638 1 gal. Alcoholic Eosin Y Stain
22-050-916 2.5 gal. Alcoholic Eosin Y Stain
22-050-197 1 pt. Eosin Phloxine Stain
22-050-198 1 gal. Eosin Phloxine Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-110-687 Supplier Epredia™ Supplier No. 71504
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated.
  • Strict quality control of raw materials and finished product ensures a high quality product every time
  • Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
  • Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
  • Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
Specifications
Quantity 4 x 1 pt.
Product Type Alcoholic Eosin Y Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov