Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
GSA_VA
Catalog No. 22050198 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
4 x 1 pt.
Product Type:
Alcoholic Eosin Y Stain
Eosin Phloxine Stain
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-198 1 gal. Eosin Phloxine Stain
22-110-637 1 pt. Alcoholic Eosin Y Stain
22-110-638 1 gal. Alcoholic Eosin Y Stain
22-050-916 2.5 gal. Alcoholic Eosin Y Stain
22-050-197 1 pt. Eosin Phloxine Stain
22-110-687 4 x 1 pt. Alcoholic Eosin Y Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-050-198 Supplier Epredia™ Supplier No. 71311
Add to Cart
Add to Cart
This item is not returnable. View return policy
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated.
  • Strict quality control of raw materials and finished product ensures a high quality product every time
  • Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
  • Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
  • Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 gal.
Product Type Eosin Phloxine Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov