Learn More
Epredia™ Richard-Allan Scientific™ Gomori's Trichrome (Blue Collagen)
Catalog No. 23900662 Shop All Epredia Products
Change view
Click to view available options
Gomori Trichrome (Blue Collagen) stain kit
Catalog No.
23-900-662 Gomori Trichrome (Blue Collagen) stain kit
1 options
Catalog No. 23-900-662 Supplier Epredia™ Supplier No. 87020
Add to Cart
Add to Cart
Save time with Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
  • Stains collagen blue, cytoplasm and muscle fibers red and the nuclei blue-to-black
Kit includes 500 mL each of:
  • Weigert's Iron Hematoxylin part A (88028)
  • Weigert's Iron Hematoxylin part B (88029)
  • Gomori's Trichrome stain solution, Blue Collagen (88030)
  • Bouin's Fluid (88038)
  • 1% Acetic Acid solution (88039)
Specifications
Product Type Stain Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov