Learn More
Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Green Collagen), Gomori Trichrome Stain Solution, Green Collagen
Catalog No. 23900663 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Gomori's Trichrome (Green Collagen) Stain Kit
Catalog No. Product Type
23-900-663 Gomori's Trichrome (Green Collagen) Stain Kit
1 options
Catalog No. 23-900-663 Supplier Epredia™ Supplier No. 87021
Add to Cart
Add to Cart
Save time with Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Green Collagen), a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
Specifications
Product Type Gomori's Trichrome (Green Collagen) Stain Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov