Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ High-Quality Dehydrants
GSA_VA

Catalog No. 22050131 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1 gal., 4/case
5 gal.
Product Type:
Acetone
Ethyl Alcohol
Isopropyl Alcohol
Methyl Alcohol
Reagent Alcohol
Concentration:
100%
70%
92%
99%
IPA:5.5%, EtOH:90%, MeOH:4.5%
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Concentration
22-050-131 1 gal., 4/case Acetone 100%
22-050-132 1 gal., 4/case Ethyl Alcohol 92%
9570 1 gal., 4/case Isopropyl Alcohol 70%
22-050-134 1 gal., 4/case Isopropyl Alcohol 99%
95-15 5 gal. Isopropyl Alcohol 99%
22-050-135 1 gal., 4/case Methyl Alcohol 99%
9990520 1 gal. Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
91-11 1 gal., 4/case Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-050-131 Supplier Epredia™ Supplier No. 9011
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

  • Formulated to rigid specifications,providing laboratories with high-quality products
  • Packaged in high-density polyethylene, which is safer and lighter than glass

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Boiling Point 56°C
Color Colorless
Compatibility Incompatible Products: Heat, flames and sparks
Content And Storage Keep containers tightly closed in a dry, cool and well-ventilated place. Keep away from heatand sources of ignition. Flammables area. Keep containers tightly closed in a cool, well ventilatedplace. Keep in properly labeled containers.
Concentration 100%
Melting Point -95°C
Product Type Acetone
Quantity 1 gal., 4/case
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.