Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ High-Quality Dehydrants

Catalog No. 22050132 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1 gal., 4/case
5 gal.
Product Type:
Acetone
Ethyl Alcohol
Isopropyl Alcohol
Methyl Alcohol
Reagent Alcohol
Concentration:
100%
70%
92%
99%
IPA:5.5%, EtOH:90%, MeOH:4.5%
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Concentration
22-050-132 1 gal., 4/case Ethyl Alcohol 92%
22-050-131 1 gal., 4/case Acetone 100%
9570 1 gal., 4/case Isopropyl Alcohol 70%
22-050-134 1 gal., 4/case Isopropyl Alcohol 99%
95-15 5 gal. Isopropyl Alcohol 99%
22-050-135 1 gal., 4/case Methyl Alcohol 99%
9990520 1 gal. Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
91-11 1 gal., 4/case Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-050-132 Supplier Epredia™ Supplier No. 9211
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

  • Formulated to rigid specifications,providing laboratories with high-quality products
  • Packaged in high-density polyethylene, which is safer and lighter than glass

Specifications

Boiling Point 77.8°C
Color Colorless
Compatibility Incompatible Materials: Strong oxidizing agents, Strong acids, alkaline
Content And Storage Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Use only outdoors or in a well-ventilated area.
Concentration 92%
Melting Point -113.3°C
Product Type Ethyl Alcohol
Quantity 1 gal., 4/case
Form Liquid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.