Learn More
Description
Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.
- Formulated to rigid specifications,providing laboratories with high-quality products
- Packaged in high-density polyethylene, which is safer and lighter than glass
Specifications
Specifications
| Boiling Point | 78.5 °C / 173.3 °F |
| Color | Colorless |
| Compatibility | Incompatible Materials: Strong oxidizing agents, Peroxides, Acids, Heavy metals |
| Content And Storage | Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Use only outdoors or in a well-ventilated area. |
| Concentration | IPA:5.5%, EtOH:90%, MeOH:4.5% |
| Melting Point | -114.1°C |
| Product Type | Reagent Alcohol |
| Quantity | 1 gal., 4/case |
| Form | Liquid |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.