Learn More
Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution
Catalog No. 88013 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Hydrochloric Acid Solution (20%)
Iron Stain Kit
Potassium Ferrocyanide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
88-013 Potassium Ferrocyanide
22-110-644 Iron Stain Kit
22-110-650 Hydrochloric Acid Solution (20%)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 88-013 Supplier Epredia™ Supplier No. 88013
Add to Cart
Edge
Add to Cart
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.
  • Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
  • Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
Specifications
Product Type Potassium Ferrocyanide