Learn More
Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y
GSA_VA
Catalog No. 23900658 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125mL
Catalog No. Quantity
23-900-658 125mL
1 options
This item is not returnable. View return policy
Catalog No. 23-900-658 Supplier Epredia™ Supplier No. 87011
Add to Cart
Add to Cart
This item is not returnable. View return policy
Effectively identify plasma cells and RNA in tissue sections and cytological preparations with Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y Stain
  • DNA is stained green
  • RNA is stained red
  • Concentrated MGP is diluted prior to use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 125mL
Product Type Methyl Green-Pyronin Y