Learn More
Epredia™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Chromic Acid) Kit
Catalog No. 88041 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 Kit
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-041 500 mL
23-900-665 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-041 Supplier Epredia™ Supplier No. 88041
Add to Cart
Add to Cart
Uniquely modified silver stain formulation.
  • Utilizes methenamine borax capsules to make a working silver solution
  • Distinguishes fungi, basement membranes, and opportunistic organisms in tissue sections
  • Chromic acid solution enhances detection of Pneumocystis carinii, histoplasmosis, and fungus organisms; eliminates non-specific background staining
  • Entities and reticular fibers stain black against a green background
Specifications
For Use With (Application) Histological and Cytological Staining
Includes 5% Chromic Acid Solution
Quantity 500 mL
Product Type Stain Kit