Learn More
Epredia™ Modified Mayers Hematoxylin
Catalog No. 22110639 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-110-639 1 pt.
1 options
This item is not returnable. View return policy
Catalog No. 22-110-639 Supplier Epredia™ Supplier No. 72804
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Produce crisp nuclear staining with Epredia™ Richard-Allan Scientific™ Modified Mayer's Hematoxylin, a progressive nuclear stain.
  • Difficult to overstain with product
  • Requires no acid rinse and does not stain mucin
  • Recommended for use when less intense results are desired
  • An effective counterstain in immunoperoxidase and special stain procedures
Specifications
Quantity 1 pt.
Product Type Hematoxylin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov