Learn More
Epredia™ Mounting Medium
Catalog No. 22110610 Shop All Epredia Products
Change view
Click to view available options
Volume (English):
4 oz.
1 pt.
Quantity:
2/Cs.
6/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Quantity
22-110-610 4 oz. 6/Cs.
22-050-102 1 pt. 2/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-610 Supplier Epredia™ Supplier No. 4112
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.
  • Rapid drying
  • Toluene-based
  • Optically clear
Ordering Alerts: Packaged in two configurations for convenience: a 1-pt. regular bottle or a 4-oz. plastic squeeze bottle with a twist-open/twist-close cap.
Specifications
Volume (English) 4 oz.
Quantity 6/Cs.
Product Type Mounting Medium
WARNING: Reproductive Harm - www.P65Warnings.ca.gov