Learn More
Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS), Schiff Reagent
GSA_VA
Catalog No. 22110645 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Hematoxylin I
Periodic Acid Schiff
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-110-645 Periodic Acid Schiff
88-018 Hematoxylin I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Alternative option to part number 860014
Catalog No. 22-110-645 Supplier Epredia™ Supplier No. 87007
Add to Cart
Edge
Add to Cart
Alternative option to part number 860014
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution
  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Periodic Acid Schiff
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov