Learn More
Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS), Schiff Reagent
Catalog No. 88018 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Hematoxylin I
Periodic Acid Schiff
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
88-018 Hematoxylin I
22-110-645 Periodic Acid Schiff
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 88-018 Supplier Epredia™ Supplier No. 88018
Add to Cart
Add to Cart
This item is not returnable. View return policy
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution
  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
Specifications
Product Type Hematoxylin I
WARNING: Reproductive Harm - www.P65Warnings.ca.gov