Learn More
Epredia™ Richard-Allan Scientific™ Reticulocyte Stain
Catalog No. 22110655 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
Catalog No. Quantity
22-110-655 125 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-110-655 Supplier Epredia™ Supplier No. 89029
Add to Cart
Add to Cart
This item is not returnable. View return policy
Identify immature erythrocytes withEpredia™ Richard-Allan Scientific™ Reticulocyte Stain
  • Immature erythrocytes stain blue
  • Mature erythrocytes do not stain
Specifications
Quantity 125 mL
Product Type Reticulocyte stain