Learn More
Epredia™ Richard-Allan Scientific™ Saccomanno Fluid
Catalog No. 22110641 Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-110-641 60 mL
22-050-215 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-641 Supplier Epredia™ Supplier No. 76160
Add to Cart
Add to Cart
This item is not returnable. View return policy
Store specimens for weeks with Epredia™ Richard-Allan Scientific™ Saccomanno Fluid, which acts as an excellent general cytological fixative.
  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
Specifications
Product Type Fixative
Quantity 60 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov