Learn More
Epredia™ Signature Series™ Pen-Fix™
Catalog No. 22046340 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 5 L
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-046340 4 x 1 gal.
22-050-316 5 gal.
61-15 4 x 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-046340 Supplier Epredia™ Supplier No. 6101
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
Specifications
Quantity 4 x 1 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov