Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Pen-Fix™

Catalog No. 6115 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 5 L
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-15 4 x 5 L
22-046340 4 x 1 gal.
22-050-316 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 61-15 Supplier Epredia™ Supplier No. 6115
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.

  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative

Specifications

Quantity 4 x 5 L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.