Learn More
Epredia™ Richard-Allan Scientific™ Specimen Transportation System (STS™ ), Containers, Prefilled; Neutral Buffered Formalin, Polypropylene
Catalog No. 22045409 Shop All Siemens Healthineers Products
Change view
Click to view available options
Containers, Prefilled; Neutral Buffered Formalin; Polypropylene; 7mL
Catalog No.
22-045-409 Containers, Prefilled; Neutral Buffered Formalin; Polypropylene; 7mL
1 options
This item is not returnable. View return policy
Catalog No. 22-045-409 Supplier Epredia™ Supplier No. 11561341
Add to Cart
Add to Cart
This item is not returnable. View return policy
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Epredia™ Richard-Allan Scientific™ Specimen Transportation System (STS™).
Closure System
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
Pre-Filled Containers
  • Nonsterile containers marked with a lot code and expiration date
  • Comply with all updated federal and state OSHA requirement
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Ordering Information
  • Clear polystyrene containers have screw-top lids
  • Opaque polyethylene containers have snap-top lids
  • Dry containers available separately