Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
GSA_VA
Catalog No. 22050275 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Epredia Three Step Stain Kit
Fixative
Solution A
Solution B
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-050-275 Solution B
22-050-272 Epredia Three Step Stain Kit
22-050-273 Fixative
22-050-274 Solution A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-050-275 Supplier Epredia™ Supplier No. 3323
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Solution B
WARNING: Reproductive Harm - www.P65Warnings.ca.gov