Learn More
Epredia™ Tissue Section Adhesive
Catalog No. 86014 Shop All Epredia Products
Change view
Click to view available options
Capacity:
500 mL
Catalog No. Capacity
86-014 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 86-014 Supplier Epredia™ Supplier No. 86014
This item is not returnable. View return policy
Increase the affinity of the tissue for the slide with Epredia™ Tissue Section Adhesive: Pre-mixed and ready-to-use.
Use in the water bath to increase the affinity of the tissue for the slide.
  • Creates no background residue
  • Improves adhesion for techniques utilizing heat
Specifications
Capacity 500 mL
Product Type Tissue Section Adhesive
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov