Learn More
Epredia™ Xylene
GSA_VA
Catalog No. 22050283 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 Gallon, Ea.
1 Gallon; 4/Cs.
5 liter; 4/Cs.
55 gallon; steel drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-283 1 Gallon; 4/Cs.
66-15 5 liter; 4/Cs.
22-110-618 55 gallon; steel drum
99-905-01 1 Gallon, Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Alternative option to part number ES609
Catalog No. 22-050-283 Supplier Epredia™ Supplier No. 6601
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Alternative option to part number ES609
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with this Xylene.
  • For use in all tissue processors and automatic stainers
  • Contains the lowest level of benzene, a known carcinogen
  • Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 Gallon; 4/Cs.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov