Learn More
Electron Microscopy Sciences Richter Anti-Vibration Pad, 7" x 8.5"
SDP
Catalog No. 5033524
Encompass
This anti-vibration pad inexpensively dampens vibrations from lab equipment. It creates a stable surface that will keep equipment in place. Fits a variety og bench-top vortexes, shakers, rockers, and microscopes. Small, lightweight design makes transportation easy. Pads can be stacked fro heavier units or laid side-by-side for larger equipment.
Size: 7" x 8½"
Catalog No. 50-335-24 Supplier Electron Microscopy Sciences Supplier No. 6711810
This anti-vibration pad inexpensively dampens vibrations from lab equipment. It creates a stable surface that will keep equipment in place. Fits a variety og bench-top vortexes, shakers, rockers, and microscopes. Small, lightweight design makes transportation easy. Pads can be stacked fro heavier units or laid side-by-side for larger equipment.
Size: 7" x 8½"