Learn More
Cayman Chemical Ricinelaidic Acid, Purity: ≥95%, CAS: 540-12-5, 250 mg
SDP
Catalog No. 502963982
Encompass_Preferred
An LTB4 receptor antagonist (Ki = 2 µM); inhibits chemotaxis and calcium flux by LTB4 in isolated human neutrophils (IC50s = 10 and 7 µM, respectively); inhibits bronchoconstriction induced by LTB4 in rats by 46%
Catalog No. 50-296-3982 Supplier Cayman Chemical Supplier No. 22470250 MG
An LTB4 receptor antagonist (Ki = 2 µM); inhibits chemotaxis and calcium flux by LTB4 in isolated human neutrophils (IC50s = 10 and 7 µM, respectively); inhibits bronchoconstriction induced by LTB4 in rats by 46%