Learn More
Medchemexpress LLC Ricinoleic acid | 141-22-0 | 5 MG
SDP
Catalog No. 5000421416
Encompass_Preferred
Ricinoleic acid (purity≥99%), a hydroxy fatty acid, is an attractive feedstock for the production of high-performance lubricants, cosmetics, polymers, surfactants, and coatings. It is the main active ingredient of Castor oil and acts as an agonist for the prostaglandin EP3 receptor, which causes laxative effects and uterine contraction. Additionally, it exhibits antianxiety-like, anti-inflammatory, and pro-inflammatory properties.
  • Acts as an agonist for prostaglandin EP3 receptor.
  • Causes laxative effects and uterine contraction.
  • Exhibits antianxiety-like properties.
  • Demonstrates anti-inflammatory and pro-inflammatory properties.
  • Can inhibit the release of β-HEX in RBL-2H3 cells.
Catalog No. 50-004-21416 Supplier Medchemexpress LLC Supplier No. HYN6070A5MG
Add to Cart
Add to Cart
Ricinoleic acid (purity≥99%), a hydroxy fatty acid, is an attractive feedstock for the production of high-performance lubricants, cosmetics, polymers, surfactants, and coatings. It is the main active ingredient of Castor oil and acts as an agonist for the prostaglandin EP3 receptor, which causes laxative effects and uterine contraction. Additionally, it exhibits antianxiety-like, anti-inflammatory, and pro-inflammatory properties.
  • Acts as an agonist for prostaglandin EP3 receptor.
  • Causes laxative effects and uterine contraction.
  • Exhibits antianxiety-like properties.
  • Demonstrates anti-inflammatory and pro-inflammatory properties.
  • Can inhibit the release of β-HEX in RBL-2H3 cells.